Learn More
Rp Cargille Laboratories Inc Cargille Immersion Liquid, Code 1160, Mixed Phthalate Esters, 30 mL
SDP
Catalog No. NC3934452
Encompass_Preferred
This multi-temperature immersion liquid is a specialized optical fluid designed for refractive index matching, primarily used with BK7 Schott glass in applications such as prisms and slides. Formulated as a colorless liquid consisting of mixed phthalate esters, it is intended for professional and research laboratory use.
  • Provides a refractive index of 1.587 at 25°C
  • Features a kinematic viscosity of 41 mm²/s at 25°C
  • Operates within a temperature range of 4°C to 40°C
  • Boasts high thermal stability with a boiling point exceeding 370°C
  • Demonstrates material compatibility with aluminum, brass, copper, nylon, and polyethylene
  • Offers an adjustment accuracy of ±0.0002
Catalog No. NC3934452 Supplier Rp Cargille Laboratories Inc Supplier No. 1934030ML
This multi-temperature immersion liquid is a specialized optical fluid designed for refractive index matching, primarily used with BK7 Schott glass in applications such as prisms and slides. Formulated as a colorless liquid consisting of mixed phthalate esters, it is intended for professional and research laboratory use.
  • Provides a refractive index of 1.587 at 25°C
  • Features a kinematic viscosity of 41 mm²/s at 25°C
  • Operates within a temperature range of 4°C to 40°C
  • Boasts high thermal stability with a boiling point exceeding 370°C
  • Demonstrates material compatibility with aluminum, brass, copper, nylon, and polyethylene
  • Offers an adjustment accuracy of ±0.0002