Learn More
Electron Microscopy Sciences Meltmount™ Slide Mounting Media 1.605 1 OZ
SDP
Catalog No. 5027100
Encompass_Preferred
Cargille Meltmount (optical quality) is a series of mounting media which are specifically formulated for use in microscope slide mounting and in other optical coupling applications.
< Use Meltmount for instant thermal mountants that require "no-oven-time" and are thermally reversible for sample retrieval or re-orientation. Meltmounts are PBC free and contains no solvents. Cleanup in toluene. Fluid at 65 degree C. It is a direct replacement for old mountant mediums (Canada Balsam, Aroclor 5442, Naphrax...) It has a refractive index (nD @25 deg C) of 1.605 and an Abbe V dispersion of 30, making it appropriate for mounting asbestiform Tremolite, Anthophyllite, and Actinolite.
Refractive index: 1.605
Catalog No. 50-271-00 Supplier Electron Microscopy Sciences Supplier No. 1799440
Cargille Meltmount (optical quality) is a series of mounting media which are specifically formulated for use in microscope slide mounting and in other optical coupling applications.
< Use Meltmount for instant thermal mountants that require "no-oven-time" and are thermally reversible for sample retrieval or re-orientation. Meltmounts are PBC free and contains no solvents. Cleanup in toluene. Fluid at 65 degree C. It is a direct replacement for old mountant mediums (Canada Balsam, Aroclor 5442, Naphrax...) It has a refractive index (nD @25 deg C) of 1.605 and an Abbe V dispersion of 30, making it appropriate for mounting asbestiform Tremolite, Anthophyllite, and Actinolite.
Refractive index: 1.605