Learn More
Medchemexpress LLC Carglumic Acid (Standard) | 1188-38-1 | 99.9% | 100 MG
SDP
Catalog No. 5000408147
Encompass_Preferred
Carglumic Acid (Standard) is an analytical standard designed for research and analytical applications. It serves as a functional analogue of N-acetylglutamate (NAG) and acts as a carbamoyl phosphate synthetase 1 (CPS1) activator. This compound is notably used in addressing acute and chronic hyperammonemia linked to NAG synthase (NAGS) deficiency.
  • Intended for research and analytical applications
  • Functional analogue of N-acetylglutamate
  • Carbamoyl phosphate synthetase 1 activator
  • Used to treat acute and chronic hyperammonemia associated with NAG synthase deficiency
  • Commonly employed in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-08147 Supplier Medchemexpress LLC Supplier No. HYB0711R100MG
Carglumic Acid (Standard) is an analytical standard designed for research and analytical applications. It serves as a functional analogue of N-acetylglutamate (NAG) and acts as a carbamoyl phosphate synthetase 1 (CPS1) activator. This compound is notably used in addressing acute and chronic hyperammonemia linked to NAG synthase (NAGS) deficiency.
  • Intended for research and analytical applications
  • Functional analogue of N-acetylglutamate
  • Carbamoyl phosphate synthetase 1 activator
  • Used to treat acute and chronic hyperammonemia associated with NAG synthase deficiency
  • Commonly employed in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS