Learn More
Medchemexpress LLC Carglumic Acid (Standard) | 1188-38-1 | 99.9% | 50 MG
SDP
Catalog No. 5000405544
Encompass_Preferred
Carglumic acid is an analytical standard and functional analogue of N-acetylglutamate (NAG), which activates carbamoyl phosphate synthetase 1 (CPS1). It is used to address acute and chronic hyperammonemia resulting from NAG synthase (NAGS) deficiency. This product is suitable for research and analytical purposes.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
  • High purity.
  • Available in solid form with a white to off-white appearance.
Catalog No. 50-004-05544 Supplier Medchemexpress LLC Supplier No. HYB0711R50MG
Carglumic acid is an analytical standard and functional analogue of N-acetylglutamate (NAG), which activates carbamoyl phosphate synthetase 1 (CPS1). It is used to address acute and chronic hyperammonemia resulting from NAG synthase (NAGS) deficiency. This product is suitable for research and analytical purposes.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
  • High purity.
  • Available in solid form with a white to off-white appearance.