Learn More
Medchemexpress LLC HY-B0711 500mg Medchemexpress, Carglumic Acid N-Carbamyl-L-glutamic acid CAS:1188-38-1 Purity:98%
SDP
Catalog No. 501639584
Encompass_Preferred
Medchemexpress,HY-B0711 500mg Carglumic Acid N-Carbamyl-L-glutamic acid CAS:1188-38-1 Formula:C6H10N2O5 Carglumic acid, a functional analogue of N-acetylglutamate (NAG) and a carbamoyl phosphate synthetase 1 (CPS1) activator, is used to treat acute and chronic hyperammonemia associated with NAG synthase (NAGS) deficiency. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9584 Supplier Medchemexpress LLC Supplier No. HYB0711500MG
Medchemexpress,HY-B0711 500mg Carglumic Acid N-Carbamyl-L-glutamic acid CAS:1188-38-1 Formula:C6H10N2O5 Carglumic acid, a functional analogue of N-acetylglutamate (NAG) and a carbamoyl phosphate synthetase 1 (CPS1) activator, is used to treat acute and chronic hyperammonemia associated with NAG synthase (NAGS) deficiency. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.