Learn More
Sigma Aldrich Fine Chemicals Biosciences Cargo interlocking manual
Catalog No. 502901332
Encompass
Ideal for your serological pipettes The Cargo manual pipette rack fulfills your laboratory needs. Click together horizontally with side connections. Securely lock in place with top connections. Magnets on both sides of units connect securely and safely to metal surface such as file cabinets fume hoods refrigerators and freezers. Angled shelves store your 0.1 to 50 mL pipettes within arms reach for use. Protective lid encloses the unit to prevent dust build up. The lid also provides an additional shelf for extra space when connected horizontally. Varying shades of blue brings diversity to the lab.
Catalog No. 50-290-1332 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6929643EA
Ideal for your serological pipettes The Cargo manual pipette rack fulfills your laboratory needs. Click together horizontally with side connections. Securely lock in place with top connections. Magnets on both sides of units connect securely and safely to metal surface such as file cabinets fume hoods refrigerators and freezers. Angled shelves store your 0.1 to 50 mL pipettes within arms reach for use. Protective lid encloses the unit to prevent dust build up. The lid also provides an additional shelf for extra space when connected horizontally. Varying shades of blue brings diversity to the lab.