Learn More
Fisherbrand™ Cargo Pipette Rack
Catalog No. 14127664 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue/Clear
Catalog No. Color
14-127-664 Blue/Clear
1 options
Catalog No. 14-127-664
Add to Cart
Add to Cart
Pipette rack designed to meet the needs of most labs.
  • Racks securely lock together horizontally or vertically to desired configuration or can be used individually
  • Can be used on top of benchtop or conveniently hung on any metallic surface with strong magnets that are designed into the racks
  • Angled shelves store 0.1 to 50 mL pipettes
  • Protective lid encloses the unit to prevent dust build-up and provides additional shelf space
  • Units lock securely together horizontally or vertically
  • Magnets connect securely to most metallic surfaces
Specifications
Autoclavable Not Autoclavable
Color Blue/Clear
Holds Pipets
Material ABS
Positions 3
Stackable Yes, Interlocking
Sterility Non-sterile
No. per Pack 3 Pk.
Dimensions (L x W x H) 11.9 x 3.9 x 5 in.
Product Type Pipet Holder