Learn More
Pyramex™ Carhartt™ Carbondale™ Safety Eyewear
Catalog No. 19155242 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Description:
Capture Clam
Polybag
Frame Color:
Black, Tan
Lens Tint:
Bronze
Clear
Gray
Mirror
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Frame Color Lens Tint
19-155-242 Capture Clam Black, Tan Clear
19-155-245 Capture Clam Black, Tan Bronze
19-155-247 Capture Clam Black, Tan Gray
19-155-250 Capture Clam Black, Tan Mirror
19-155-249 Polybag Black, Tan Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-155-242 Supplier Pyramex™ Supplier No. CHB210DCC
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rugged wraparound safety glasses for any job site
  • Flexible rubber nosepiece
  • Co-injected temples
  • Available in a Carhartt capture clam or polybag
  • Suspended anti-fog lens option available
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Description Capture Clam
    Lens Tint Clear
    Frame Color Black, Tan
    Lens Material Polycarbonate
    Frame Material Nylon