Learn More
Pyramex™ Carhartt™ Cayce™ Safety Eyewear
Catalog No. 19155274 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Bronze
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-274 Clear
19-155-275 Bronze
19-155-276 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-274 Supplier Pyramex™ Supplier No. CHB910ST
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
All lenses provide 99% protection from harmful UV rays and are hard coated polycarbonate, scratch-resistant
  • Adjustable rubber nosepiece
  • Slim temple design
  • Wrap around coverage for superior protection
  • Available in a Carhartt polybag
  • Specifications
    Certifications/Compliance ANSI Z87.1+,CSA Z94.3
    Lens Tint Clear
    Frame Color Black
    Lens Material Polycarbonate
    Frame Material Polycarbonate