Learn More
Pyramex™ Carhartt™ Easley™ Safety Eyewear
Catalog No. 19155273 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Bronze
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-273 Gray
19-155-271 Clear
19-155-272 Bronze
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-273 Supplier Pyramex™ Supplier No. CHB820ST
Add to Cart
Add to Cart
This item is not returnable. View return policy
Antifog lens in polybag
  • Built-in browguard
  • Adjustable nosepiece
  • Ventilated temples
  • Available in a Carhartt polybag
  • Specifications
    Certifications/Compliance ANSI Z87.1+,CSA Z94.3
    Lens Tint Gray
    Frame Color Black, Tan
    Weight (Metric) 31 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate