Learn More
Pyramex™ Carhartt™ Ironside™ Safety Eyewear
Catalog No. 19155261 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Description:
Capture Clam
Polybag
Frame Color:
Black, Tan
Lens Tint:
Bronze
Gray
Mirror
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Frame Color Lens Tint
19-155-261 Capture Clam Black, Tan Bronze
19-155-263 Capture Clam Black, Tan Gray
19-155-265 Capture Clam Black, Tan Mirror
19-155-260 Polybag Black, Tan Bronze
19-155-262 Polybag Black, Tan Gray
19-155-264 Polybag Black, Tan Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 19-155-261 Supplier Pyramex™ missing translation for 'supplierNo' CHB618DTCC
Only null left
Add to cart
Add to cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Lenses provide 99% protection from harmful UV rays and are hard coated polycarbonate, scratch-resistant. Meets ANSI Z87.1 High Impact Requirements.
  • Straight-back vented, flexible temples
  • Rubber nosepiece
  • Rubber temple tips
  • Available in a Carhartt capture clam or polybag
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Description Capture Clam
    Lens Tint Bronze
    Frame Color Black, Tan
    Weight (Metric) 30.6 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate