Learn More
Pyramex™ Carhartt™ Spokane™ Safety Eyewear
Catalog No. 19155252 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Bronze
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-252 Clear
19-155-254 Bronze
19-155-256 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-252 Supplier Pyramex™ Supplier No. CHB310DCC
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Half-frame safety glasses for everyday work wear
  • Flexible rubber nosepiece
  • Co-injected straight-back temples
  • Unique half-frame design for the active lifestyle
  • Available in a Carhartt capture clam or polybag
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Description Capture Clam
    Lens Tint Clear
    Frame Color Black
    Weight (Metric) 24 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate