Learn More
Pyramex™ Carhartt™ Spokane™ Safety Eyewear
Catalog No. 19155256 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Bronze
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-256 Gray
19-155-252 Clear
19-155-254 Bronze
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-256 Supplier Pyramex™ Supplier No. CHB320DCC
Add to Cart
Add to Cart
This item is not returnable. View return policy
Half-frame safety glasses for everyday work wear
  • Flexible rubber nosepiece
  • Co-injected straight-back temples
  • Unique half-frame design for the active lifestyle
  • Available in a Carhartt capture clam or polybag
  • Specifications
    Lens Coating Scratch-resistant
    Certifications/Compliance ANSI Z87.1
    Description Capture Clam
    Lens Tint Gray
    Product Type Safety Glasses
    Frame Color Black
    Weight (Metric) 24 g
    Material Polycarbonate, TPR
    Disposable Reusable
    Lens Material Polycarbonate
    Frame Material Polycarbonate
    UV Protection 99.9% UVA/UVB/UVC
    Frame Type Half Frame