Learn More
Medchemexpress LLC HY-14763 5mg Medchemexpress, Cariprazine CAS:839712-12-8 Purity:>98%
SDP
Catalog No. 502030554
Encompass_Preferred
Medchemexpress, HY-14763 5mg Cariprazine CAS:839712-12-8 Cariprazine is a novel antipsychotic drug candidate that exhibits high affinity for the D3 (Ki=0.085 nM) and D2 (Ki=0.49 nM) receptors, and moderate affinity for the 5-HT1A receptor (Ki=2.6 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0554 Supplier Medchemexpress LLC Supplier No. HY147635MG
Medchemexpress, HY-14763 5mg Cariprazine CAS:839712-12-8 Cariprazine is a novel antipsychotic drug candidate that exhibits high affinity for the D3 (Ki=0.085 nM) and D2 (Ki=0.49 nM) receptors, and moderate affinity for the 5-HT1A receptor (Ki=2.6 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.