Learn More
Medchemexpress LLC Cariprazine-d6 | 1308278-67-2 | MFCD00038264 | 98.7% | 433.45 | C21H26D6Cl2N4O | 10 MG
SDP
Catalog No. 5000286969
Encompass_Preferred
Cariprazine-d6 is a deuterium-labeled analogue of the antipsychotic cariprazine provided as a characterized reference standard for research use. It contains six deuterium atoms and is intended for use as an internal standard or tracer in analytical, pharmacokinetic, and metabolite studies; supporting documentation includes a product data sheet and safety data sheet.
Catalog No. 50-002-86969 Supplier Medchemexpress LLC Supplier No. HY14763S10MG
Cariprazine-d6 is a deuterium-labeled analogue of the antipsychotic cariprazine provided as a characterized reference standard for research use. It contains six deuterium atoms and is intended for use as an internal standard or tracer in analytical, pharmacokinetic, and metabolite studies; supporting documentation includes a product data sheet and safety data sheet.