Learn More
Medchemexpress LLC Cariprazine-d8 | 1308278-50-3 | 98.8% | 435.46 | 1 MG
SDP
Catalog No. 5000360561
Encompass_Preferred
Cariprazine-d8 is a deuterium labeled Cariprazine, a novel antipsychotic agent candidate exhibiting high affinity for the D3 (Ki=0.085 nM) and D2 (Ki=0.49 nM) receptors, and moderate affinity for the 5-HT1A receptor (Ki=2.6 nM).
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-60561 Supplier Medchemexpress LLC Supplier No. HY14763S11MG
Cariprazine-d8 is a deuterium labeled Cariprazine, a novel antipsychotic agent candidate exhibiting high affinity for the D3 (Ki=0.085 nM) and D2 (Ki=0.49 nM) receptors, and moderate affinity for the 5-HT1A receptor (Ki=2.6 nM).
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS