Learn More
Medchemexpress LLC Cariprazine-d8 | 1308278-50-3 | 99.5% | 435.46 | C21H24D8Cl2N4O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14763S110MG
Encompass_Preferred
Cariprazine-d8 is a deuterium-labeled analogue of the antipsychotic candidate cariprazine, supplied as a solid analytical reference standard for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS assays. It is provided as a high-purity white to off-white solid with recommended sealed storage to maintain stability.
  • Deuterium-labeled internal standard for accurate quantitation
  • High purity (99.51%) for reliable analytical results
  • Suitable for NMR, GC-MS, and LC-MS applications
  • White to off-white solid for easy handling and visual QC
  • Recommended sealed storage to preserve stability
  • Controlled-substance restrictions may apply in some territories
Catalog No. 50-002-87658