Learn More
Medchemexpress LLC Cariprazine hydrochloride | 1083076-69-0 | 99.71% | 463.87 | 1 ML
SDP
Catalog No. 5000362535
Encompass_Preferred
Cariprazine hydrochloride is a novel antipsychotic agent candidate that exhibits high affinity for D3 (Ki=0.085 nM) and D2 (Ki=0.49 nM) receptors, and moderate affinity for the 5-HT1A receptor (Ki=2.6 nM). It is currently being investigated in various clinical trials for conditions such as Bipolar Disorder, Schizophrenia, and Major Depressive Disorder.
  • High affinity for D3 (0.085 nM) and D2 (0.49 nM) receptors
  • Moderate affinity for 5-HT1A receptor (2.6 nM)
  • Antimanic properties
  • Interferes with dopamine D2 receptor β-arrestin interactions
  • Reduces ouabain-induced hyperactivity
  • Available as a solution in DMSO
Catalog No. 50-003-62535 Supplier Medchemexpress LLC Supplier No. HY14763A10MM/1ML
Cariprazine hydrochloride is a novel antipsychotic agent candidate that exhibits high affinity for D3 (Ki=0.085 nM) and D2 (Ki=0.49 nM) receptors, and moderate affinity for the 5-HT1A receptor (Ki=2.6 nM). It is currently being investigated in various clinical trials for conditions such as Bipolar Disorder, Schizophrenia, and Major Depressive Disorder.
  • High affinity for D3 (0.085 nM) and D2 (0.49 nM) receptors
  • Moderate affinity for 5-HT1A receptor (2.6 nM)
  • Antimanic properties
  • Interferes with dopamine D2 receptor β-arrestin interactions
  • Reduces ouabain-induced hyperactivity
  • Available as a solution in DMSO