Learn More
Selleck Chemical LLC Cariprazine hydrochloride
SDP
Catalog No. 5000000850
Encompass_Preferred
Cariprazine hydrochloride(RGH188 hydrochloride) is a novel antipsychotic agent and is a highly selective partial agonist of dopamineD2 and D3 and 5HT1A receptors with preferential binding to the D3 receptor It exhibits high selectivity and affinity to dopamine D3 (Ki 0 085 nM) and D2 (Ki 0 49 nM) receptors and has moderate affinity for the 5-HT1A receptor (Ki 2 6 nM)
Catalog No. 50-000-00850 Supplier Selleck Chemical LLC Supplier No. S723650MG
Cariprazine hydrochloride(RGH188 hydrochloride) is a novel antipsychotic agent and is a highly selective partial agonist of dopamineD2 and D3 and 5HT1A receptors with preferential binding to the D3 receptor It exhibits high selectivity and affinity to dopamine D3 (Ki 0 085 nM) and D2 (Ki 0 49 nM) receptors and has moderate affinity for the 5-HT1A receptor (Ki 2 6 nM)