Learn More
Aviva Systems Biology Corp NAXD Antibody - middle region (ARP47405_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47405_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ10769. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CARKD. Alternate names: FLJ34548, LP3298ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54000