Learn More
Sino Biological CARKL Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 14833T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CARKL (rh CARKL; Catalog#14833-H20B; NP_037408.2; Ala2-Ser478). CARKL specific IgG was purified by Human CARKL affinity chromatography.
Catalog No. 50-224-6396