Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER COULOMETRIC GENERATOR SOLUTION, PYRIDINE-FREE | 125 ML | GFS CHEMICALS, INC. Item #: 99781 | Bulk #: 1613
SDP
Catalog No. NC9759594
Encompass_Preferred
WATERMARK KARL FISCHER COULOMETRIC GENERATOR SOLUTION, PYRIDINE-FREE | For coulometric analysis. Catholyte for coulometric titrators.
Catalog No. NC9759594 Supplier GFS Chemicals Inc Supplier No. 99781
WATERMARK KARL FISCHER COULOMETRIC GENERATOR SOLUTION, PYRIDINE-FREE | For coulometric analysis. Catholyte for coulometric titrators.