Learn More
Carl Zeiss™ Axio Lab™ A1 Upright Laboratory Microscope Kits
Catalog No. 12071330
Change view
Click to view available options
Type:
Upright Microscope
Catalog No. Type
12-071-330 Upright Microscope
1 options
Catalog No. 12-071-330 Supplier Carl Zeiss™ Supplier No. 4109990000027
Axio Lab A1 Upright Laboratory Microscopes available as a dual-observation kit or hematology kit
Dual-Observation kit
  • Offers 10X/20mm Br. foc. E-PL eyepieces
  • Co-observation center piece, 40/60 L Scope
  • Filter mount for multidiscussion
  • Binoc Tube 30/23 Bio
  • Intermediate plate
Hematology kit
  • Offers 10X/20mm E-PL eyepieces
  • Right-hand mechanical stage with specimen holder
  • Five-position revolving nosepiece
  • N-Achroplan Objective 50X/1.0 oil WD
  • 0.22 M27,
  • Four A-Plan objectives (5X, 10X, 40X, 100X oil), white balance and contrast enhancement filters, and white light 3W warm LED lamp
Camera not included in the kit.
Specifications
Type Upright Microscope
For Use With (Application) Lab, educational applications