Learn More
Bioworld Carlson Lysis Buffer for CTAB DNA Extraction, 500 mL
Catalog No. 50196448
Encompass_Preferred
Carlson Lysis Buffer is a DNA purification buffer using CTAB and suitable for extraction of fungal DNA.RNAse and DNAse freeContains/ Composition:100mM Tris pH 9.52% CTAB1.4M NaCl1% PEG 6000 or 8000 (please make sure to request which when ordering)20mM EDTA Reference: Carlson JE, Tulsieram LK, Glaubitz JC, Luk VMK, Kauffeldt C, and Rutledge R (1991) Segregation of random amplified DNA markers in F1 progeny of conifers. Theor. Appl. Genet. 83:194-200. Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.
Catalog No. 50-196-448 Supplier Bioworld Supplier No. 104500022
Carlson Lysis Buffer is a DNA purification buffer using CTAB and suitable for extraction of fungal DNA.RNAse and DNAse freeContains/ Composition:100mM Tris pH 9.52% CTAB1.4M NaCl1% PEG 6000 or 8000 (please make sure to request which when ordering)20mM EDTA Reference: Carlson JE, Tulsieram LK, Glaubitz JC, Luk VMK, Kauffeldt C, and Rutledge R (1991) Segregation of random amplified DNA markers in F1 progeny of conifers. Theor. Appl. Genet. 83:194-200. Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.