Learn More
Medchemexpress LLC CARM1-IN-3 dihydrochloride | 912970-67-3 | 98.8% | 481.46 | C24H34Cl2N4O2 | 10 MG
SDP
Catalog No. 5000286585
Encompass_Preferred
CARM1-IN-3 dihydrochloride is a small-molecule inhibitor selective for co-activator associated arginine methyltransferase 1 (CARM1). Supplied as a white to off-white solid, it shows potent inhibition of CARM1 (IC50 = 0.07 μM) with minimal activity versus CARM3, and is intended for research use in biochemical and cellular assays. Store sealed and away from moisture; follow recommended cold storage for long-term stability.
  • Potent and selective CARM1 inhibition (IC50 = 0.07 μM).
  • High purity for reproducible experimental results (98.77%).
  • White to off-white solid suitable for analytical and biological applications.
  • Dihydrochloride salt form compatible with aqueous preparations.
  • Recommended storage: sealed, away from moisture; in solvent -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-86585 Supplier Medchemexpress LLC Supplier No. HY15214A10MG
CARM1-IN-3 dihydrochloride is a small-molecule inhibitor selective for co-activator associated arginine methyltransferase 1 (CARM1). Supplied as a white to off-white solid, it shows potent inhibition of CARM1 (IC50 = 0.07 μM) with minimal activity versus CARM3, and is intended for research use in biochemical and cellular assays. Store sealed and away from moisture; follow recommended cold storage for long-term stability.
  • Potent and selective CARM1 inhibition (IC50 = 0.07 μM).
  • High purity for reproducible experimental results (98.77%).
  • White to off-white solid suitable for analytical and biological applications.
  • Dihydrochloride salt form compatible with aqueous preparations.
  • Recommended storage: sealed, away from moisture; in solvent -80°C (6 months), -20°C (1 month).