Learn More
Medchemexpress LLC CARM1-IN-3 dihydrochloride | 912970-67-3 | C24H34Cl2N4O2 | 50 MG
SDP
Catalog No. 5000373596
Encompass_Preferred
CARM1-IN-3 dihydrochloride (compound 17b) is a potent and selective co-activator associated arginine methyltransferase (CARM1) inhibitor.
  • Potent and selective CARM1 inhibitor
  • IC50 value of 0.07 μM for CARM1
  • IC50 value of >25 μM for CARM3
Catalog No. 50-003-73596 Supplier Medchemexpress LLC Supplier No. HY15214A50MG
CARM1-IN-3 dihydrochloride (compound 17b) is a potent and selective co-activator associated arginine methyltransferase (CARM1) inhibitor.
  • Potent and selective CARM1 inhibitor
  • IC50 value of 0.07 μM for CARM1
  • IC50 value of >25 μM for CARM3