Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CARM1-IN-3 dihydrochloride | C24H34Cl2N4O2 | 25 MG
SDP

Catalog No. 5000374652
Encompass_Preferred

CARM1-IN-3 dihydrochloride is a potent and selective co-activator associated arginine methyltransferase (CARM1) inhibitor. It demonstrates high specificity with an IC50 of 0.07 μM for CARM1, while showing significantly less activity against CARM3 (>25 μM).

  • Potent and selective CARM1 inhibitor
  • Purity of 98.77%
  • Solid, white to off-white appearance
  • Soluble in DMSO and H2O (50 mg/mL)
  • Recommended storage at 4°C (sealed, away from moisture)

Catalog No. 50-003-74652 Supplier Medchemexpress LLC Supplier No. 25MGHY15214A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CARM1-IN-3 dihydrochloride is a potent and selective co-activator associated arginine methyltransferase (CARM1) inhibitor. It demonstrates high specificity with an IC50 of 0.07 μM for CARM1, while showing significantly less activity against CARM3 (>25 μM).

  • Potent and selective CARM1 inhibitor
  • Purity of 98.77%
  • Solid, white to off-white appearance
  • Soluble in DMSO and H2O (50 mg/mL)
  • Recommended storage at 4°C (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.