Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carm1-in-3 dihydrochloride | C24H34Cl2N4O2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY15214A5MG

Encompass_Preferred

CARM1-IN-3 dihydrochloride (compound 17b) is a potent and selective co-activator associated arginine methyltransferase (CARM1) inhibitor with IC50 values of 0.07 μM for CARM1 and >25 μM for CARM3. This compound is a solid with a white to off-white appearance.

  • Potent and selective CARM1 inhibitor
  • IC50 for CARM1: 0.07 μM
  • IC50 for CARM3: >25 μM
  • Soluble in DMSO and H2O
  • Appearance: solid, white to off-white

Catalog No. 50-003-72993


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.