Learn More
Enzo Life Sciences CARMA1 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1010100
Encompass_Preferred
Immunogen: Recombinant fragment corresponding to a region within amino acids 25 and 275 of human CARMA1. Species reactivity: Human, Mouse. Applications: IHC (PS), WB. Formulation: Liquid. In 0.1M TRIS, pH 7, containing 10% glycerol, 0.1M glycine, and 0.01% thimerosal.
Catalog No. 50-201-2662