Learn More
Sklar Instruments Surgi-OR Rochester-Carmalt Forceps, Sklar 8 in., Cross-Serrated Tips, Curved, Longitudinally Serrated, Stainless Steel. EA.
SDP
Catalog No. 501184226 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR Rochester-Carmalt Forceps, Sklar. 8 in., Cross-Serrated Tips, Curved, Longitudinally Serrated, Stainless Steel. Surgi-OR Rochester-Carmalt Hemostatic Forceps are a standard type of hemostatic clamp used in several procedures. These forceps have longitudinal serrations the entire length of the jaw. The tips are cross-serrated. The fully serrated working surface allows this instrument to have greater grip on heavy or tough tissue and stop blood flow of large vessels. The ratcheted handle allows locking on larger tissue for hemostasis. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is curved with longitudinally serrated jaws, cross-serrated tips, and a length of 8 inches.
Catalog No. 50-118-4226 Supplier Sklar Instruments Supplier No. 95542
Surgi-OR Rochester-Carmalt Forceps, Sklar. 8 in., Cross-Serrated Tips, Curved, Longitudinally Serrated, Stainless Steel. Surgi-OR Rochester-Carmalt Hemostatic Forceps are a standard type of hemostatic clamp used in several procedures. These forceps have longitudinal serrations the entire length of the jaw. The tips are cross-serrated. The fully serrated working surface allows this instrument to have greater grip on heavy or tough tissue and stop blood flow of large vessels. The ratcheted handle allows locking on larger tissue for hemostasis. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is curved with longitudinally serrated jaws, cross-serrated tips, and a length of 8 inches.