Learn More
Sklar Instruments Carmalt Splinter Forceps, Sklar, 4-1/4 in., Sharp, Straight, Serrated, Stainless Steel. EA/1.
SDP
Catalog No. 501437439 Shop All Sklar Instruments Products
Encompass_Preferred
Carmalt Splinter Forceps, Sklar, 4-1/4 in., Sharp, Straight, Serrated, Stainless Steel. Sklar Carmalt Splinter Forceps are thumb forceps commonly used in dermatological procedures. They feature an arrowhead style tip with a central groove. They may also be known as Hunter forceps. This product is straight with serrated tips, and a length of 4-1/4 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7439 Supplier Sklar Instruments Supplier No. 193240
Carmalt Splinter Forceps, Sklar, 4-1/4 in., Sharp, Straight, Serrated, Stainless Steel. Sklar Carmalt Splinter Forceps are thumb forceps commonly used in dermatological procedures. They feature an arrowhead style tip with a central groove. They may also be known as Hunter forceps. This product is straight with serrated tips, and a length of 4-1/4 inches.