Learn More
Medchemexpress LLC Carmine | 1390-65-4 | 100 MG
SDP
Catalog No. 5000415764
Encompass_Preferred
Carmine, also known as Carmine red, is a natural red dye extracted from the dried females of the Dactylopius coccus var. Costa insect, commonly known as cochineal. It is widely utilized as a food additive and can trigger both immediate hypersensitivity and delayed systemic responses with cutaneous expression.
  • Natural red dye derived from cochineal insects.
  • Widely used as a food additive.
  • Can provoke immediate hypersensitivity.
  • May cause delayed systemic responses with cutaneous expression.
  • Used in biological staining to selectively demonstrate nuclei.
  • Used in biological staining to selectively demonstrate chromosomes.
  • Used in biological staining to selectively demonstrate mucins.
Catalog No. 50-004-15764 Supplier Medchemexpress LLC Supplier No. HYN1389100MG
Carmine, also known as Carmine red, is a natural red dye extracted from the dried females of the Dactylopius coccus var. Costa insect, commonly known as cochineal. It is widely utilized as a food additive and can trigger both immediate hypersensitivity and delayed systemic responses with cutaneous expression.
  • Natural red dye derived from cochineal insects.
  • Widely used as a food additive.
  • Can provoke immediate hypersensitivity.
  • May cause delayed systemic responses with cutaneous expression.
  • Used in biological staining to selectively demonstrate nuclei.
  • Used in biological staining to selectively demonstrate chromosomes.
  • Used in biological staining to selectively demonstrate mucins.