Learn More
Electron Microscopy Sciences Carmine, Certified, C.I. DcCa-16 10 GR
SDP
Catalog No. 50980289
Encompass
(Natural Red 4)
F.W. 1150.86
Solubility: 0.0% Water; 0.2% Alcohol; 0.5% Cellosolve; 4.0% Glycol; 0.0% Xylene
Alum Lake of Carminic acid is also known as Natural Red 4.
Catalog No. 50-980-289 Supplier Electron Microscopy Sciences Supplier No. 12410
(Natural Red 4)
F.W. 1150.86
Solubility: 0.0% Water; 0.2% Alcohol; 0.5% Cellosolve; 4.0% Glycol; 0.0% Xylene
Alum Lake of Carminic acid is also known as Natural Red 4.