Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carminic Acid (Natural dye), TCI America™
SDP

Catalog No. C07825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C07825G Supplier TCI America Supplier No. C07825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Natural dye suitable for use as a laboratory reagent.

Chemical Identifiers

CAS 1260-17-9
Molecular Formula C22H20O13
Molecular Weight (g/mol) 492.39
MDL Number MFCD00167028
InChI Key DGQLVPJVXFOQEV-NGOCYOHBSA-N
Synonym carminic acid, carmine, natural red 4, ci natural red 4, san-ei gen san red 1, unii-cid8z8n95n, coccinellin, cochineal, e 120 dye, ccris 1397
PubChem CID 10255083
ChEBI CHEBI:78310
IUPAC Name 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2R,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
SMILES CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C([C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=C(O)C(O)=C1C2=O

Specifications

Quantity 5 g
Formula Weight 492.39
Percent Purity ≥80.0%
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Carminic Acid (Natural dye)
EINECSNumber (9)-0301
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.