Learn More
Selleck Chemical LLC Carmofur S1289-10mg
SDP
Catalog No. 503111421
Encompass_Preferred
Carmofur (HCFU) is a highly potent acid ceramidase inhibitor used in the treatment of breast and colorectal cancer
Catalog No. 50-311-1421 Supplier Selleck Chemical LLC Supplier No. S128910MG
Carmofur (HCFU) is a highly potent acid ceramidase inhibitor used in the treatment of breast and colorectal cancer