Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carmoisine, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014017 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4017 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4017 Supplier MilliporeSigma Supelco Supplier No. 5224525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 3567-69-9
Flash Point >225°C
Physical Form Neat
Linear Formula C20H12N2 Na2O7S2
MDL Number MFCD00003978
Synonym Chromotrope FB; Acid Red 14; Disodium 4-hydroxy-3-[(4-sulfo-1-naphthalenyl)azo]-1-naphthalenesulfonate; Mordant Blue 79
Type Analytical Standard
Quantity 25 mg
Formula Weight 502.43
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.