Learn More
Medchemexpress LLC Carmoterol hydrochloride | 137888-11-0 | 98.7% | 404.89 | C21H25ClN2O4 | 10MG
SDP
Catalog No. 5000236594
Encompass_Preferred
Carmoterol hydrochloride is a potent, selective, long-acting β2-adrenoceptor agonist supplied as a solid research chemical. It is used in pharmacological and respiratory research to study bronchoconstriction and treatments for asthma and chronic obstructive pulmonary disease (COPD), exhibiting strong bronchodilatory activity and high β2 vs β1 selectivity.
  • Potent, selective β2-adrenoceptor agonist.
  • Long-acting bronchodilator activity suitable for respiratory research.
  • High β2 vs β1 selectivity, reducing off-target cardiac β1 activation.
  • Supplied as a solid research-grade chemical for laboratory use.
  • High purity (98.7% by HPLC reported on certificate of analysis).
  • Molecular formula C21H25ClN2O4, molecular weight 404.89 g·mol-1.
Catalog No. 50-002-36594 Supplier Medchemexpress LLC Supplier No. HY1246310MG
Carmoterol hydrochloride is a potent, selective, long-acting β2-adrenoceptor agonist supplied as a solid research chemical. It is used in pharmacological and respiratory research to study bronchoconstriction and treatments for asthma and chronic obstructive pulmonary disease (COPD), exhibiting strong bronchodilatory activity and high β2 vs β1 selectivity.
  • Potent, selective β2-adrenoceptor agonist.
  • Long-acting bronchodilator activity suitable for respiratory research.
  • High β2 vs β1 selectivity, reducing off-target cardiac β1 activation.
  • Supplied as a solid research-grade chemical for laboratory use.
  • High purity (98.7% by HPLC reported on certificate of analysis).
  • Molecular formula C21H25ClN2O4, molecular weight 404.89 g·mol-1.