Learn More
Medchemexpress LLC Carmoterol hydrochloride | 137888-11-0 | 98.7% | 404.89 | C21H25ClN2O4 | 50MG
SDP
Catalog No. 5000238085
Encompass_Preferred
Carmoterol hydrochloride is a long-acting, selective β2-adrenoceptor agonist supplied for laboratory research. It is provided as a solid for studies of bronchodilation in asthma and chronic obstructive pulmonary disease (COPD) models, with supplier-provided purity and storage data to support experimental planning.
  • High purity suitable for research (98.7%).
  • Solid, white to light yellow appearance.
  • Available in multiple package sizes, including 50 MG.
  • Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Useful for in vitro and in vivo respiratory pharmacology studies.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-38085 Supplier Medchemexpress LLC Supplier No. HY1246350MG
Carmoterol hydrochloride is a long-acting, selective β2-adrenoceptor agonist supplied for laboratory research. It is provided as a solid for studies of bronchodilation in asthma and chronic obstructive pulmonary disease (COPD) models, with supplier-provided purity and storage data to support experimental planning.
  • High purity suitable for research (98.7%).
  • Solid, white to light yellow appearance.
  • Available in multiple package sizes, including 50 MG.
  • Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Useful for in vitro and in vivo respiratory pharmacology studies.
  • For research use only; not for human or clinical use.