Learn More
Medchemexpress LLC HY-103410 5mg Medchemexpress, Carmoxirole (hydrochloride) CAS:115092-85-8 Purity:>98%
SDP
Catalog No. 501968104
Encompass_Preferred
Medchemexpress, HY-103410 5mg Carmoxirole (HCl) CAS:115092-85-8 Carmoxirole hydrochloride (EMD 45609 hydrochloride) is a selective, peripherally acting dopamine D2 receptor agonist and exhibits antihypertensive activities in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8104 Supplier Medchemexpress LLC Supplier No. HY1034105MG
Medchemexpress, HY-103410 5mg Carmoxirole (HCl) CAS:115092-85-8 Carmoxirole hydrochloride (EMD 45609 hydrochloride) is a selective, peripherally acting dopamine D2 receptor agonist and exhibits antihypertensive activities in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.