Learn More
TARGETMOL CHEMICALS INC CARMOXIROLE HYDROCHLORID 5MG
SDP
Catalog No. 502581692
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carmoxirole hydrochloride (EMD 45609 hydrochloride) is a selective dopamine D2 receptor agonist with antihypertensive activity in vivo. purity: 98%
Catalog No. 50-258-1692 Supplier TARGETMOL CHEMICALS INC Supplier No. T106835MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carmoxirole hydrochloride (EMD 45609 hydrochloride) is a selective dopamine D2 receptor agonist with antihypertensive activity in vivo. purity: 98%