Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carmoxirole hydrochloride | 115092-85-8 | 99.8% | 410.9 | C24H27ClN2O2 | 1 ML
SDP

Catalog No. 5000244845
Encompass_Preferred

Carmoxirole hydrochloride is a research-grade small molecule described as a selective, peripherally acting dopamine D2 receptor agonist with reported antihypertensive activity in vivo.

  • Selective peripheral dopamine D2 receptor agonist.
  • Reported antihypertensive activity in vivo.
  • High purity (99.82%).
  • Available as a 10 mM solution (1 mL in DMSO) and as solid quantities for analytical use.
  • Low solubility in water; soluble in DMSO.
  • Recommended storage: solid at 4°C; in solvent, -80°C (up to 6 months) or -20°C (short term).
  • Supporting documentation includes data sheet, COA, and SDS.

Catalog No. 50-002-44845 Supplier Medchemexpress LLC Supplier No. HY10341010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Carmoxirole hydrochloride is a research-grade small molecule described as a selective, peripherally acting dopamine D2 receptor agonist with reported antihypertensive activity in vivo.

  • Selective peripheral dopamine D2 receptor agonist.
  • Reported antihypertensive activity in vivo.
  • High purity (99.82%).
  • Available as a 10 mM solution (1 mL in DMSO) and as solid quantities for analytical use.
  • Low solubility in water; soluble in DMSO.
  • Recommended storage: solid at 4°C; in solvent, -80°C (up to 6 months) or -20°C (short term).
  • Supporting documentation includes data sheet, COA, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.