Learn More
Medchemexpress LLC Carmoxirole hydrochloride | 115092-85-8 | MFCD00872970 | 99.8% | 410.94 g/mol | C24H27ClN2O2 | 100 MG
SDP
Catalog No. 5000244768
Encompass_Preferred
Carmoxirole hydrochloride is the hydrochloride salt of carmoxirole, a selective, peripherally acting dopamine D2 receptor agonist with reported antihypertensive activity in vivo. It is supplied for research use as a high-purity reference standard, available in solid form and as a DMSO solution.
  • Selective, peripherally acting dopamine D2 receptor agonist.
  • Exhibits antihypertensive activity in vivo.
  • Supplied as the hydrochloride salt for improved stability and solubility.
  • High purity (≈99.8%) suitable for research applications.
  • Available as solid in multiple milligram sizes and as a 10 mM solution in DMSO.
  • Recommended storage: 4°C for solid; in solvent, -80°C up to 6 months or -20°C up to 1 month, sealed and away from moisture.
Catalog No. 50-002-44768 Supplier Medchemexpress LLC Supplier No. HY103410100MG
Carmoxirole hydrochloride is the hydrochloride salt of carmoxirole, a selective, peripherally acting dopamine D2 receptor agonist with reported antihypertensive activity in vivo. It is supplied for research use as a high-purity reference standard, available in solid form and as a DMSO solution.
  • Selective, peripherally acting dopamine D2 receptor agonist.
  • Exhibits antihypertensive activity in vivo.
  • Supplied as the hydrochloride salt for improved stability and solubility.
  • High purity (≈99.8%) suitable for research applications.
  • Available as solid in multiple milligram sizes and as a 10 mM solution in DMSO.
  • Recommended storage: 4°C for solid; in solvent, -80°C up to 6 months or -20°C up to 1 month, sealed and away from moisture.