Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carmustine 98.0+%, TCI America™
SDP

Catalog No. C2634100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2634100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2634100MG Supplier TCI America Supplier No. C2634100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 154-93-8
Molecular Formula C5H9Cl2N3O2
Molecular Weight (g/mol) 214.05
MDL Number MFCD00057706
InChI Key DLGOEMSEDOSKAD-UHFFFAOYSA-N
Synonym 1,3-Bis(2-chloroethyl)-1-nitrosourea
PubChem CID 2578
ChEBI CHEBI:3423
IUPAC Name 1,3-bis(2-chloroethyl)-3-nitrosourea
SMILES ClCCNC(=O)N(CCCl)N=O

Specifications

Melting Point 32°C
Quantity 100 mg
UN Number 2811
Formula Weight 214.05
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Carmustine
RTECSNumber YS2625000
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.