Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Carnauba wax No. 1 yellow

Catalog No. 243213250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-321-3250G 250 g
24-321-31KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-321-3250G Supplier Sigma Aldrich Supplier No. 243213250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Carnauba wax No.1 yellow(CW) is an ester based wax that can be extracted from palm leaves. The composition of this wax contains aliphatic esters, -d-hydroxy esters and unsaturated alcohols. It can be used as a glossing agent in a variety of pharmaceutical products.CW may be used as a hydrophobic excipient which can be used to modify the kinetics of polymeric gel in a drug releasing system. It can be used in dry polymeric coatings which can increase the crystallization and improve the stability of amorphous solid drug formulations. It may also be used as a component of lipids which can be used to tune and improve the flowability and designing of solid drugs.

Specifications

CAS 8015-86-9
MDL Number MFCD00130724
Quantity 250 g
Synonym Brazil wax
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.