Learn More
Medchemexpress LLC (±)-Carnitine-d9 chloride | 1219386-75-0 | 99.7% | 5 MG
SDP
Catalog No. 5000411630
Encompass_Preferred
The product is (±)-Carnitine-d9 chloride, a deuterium-labeled form of (±)-Carnitine chloride. This compound exists in both D and L isomers, with L-carnitine known for its vital role in fatty acid β-oxidation, as well as its antioxidant and anti-inflammatory properties. This deuterated version is particularly useful as a tracer and internal standard in various analytical techniques.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11630 Supplier Medchemexpress LLC Supplier No. HYB1453S15MG
The product is (±)-Carnitine-d9 chloride, a deuterium-labeled form of (±)-Carnitine chloride. This compound exists in both D and L isomers, with L-carnitine known for its vital role in fatty acid β-oxidation, as well as its antioxidant and anti-inflammatory properties. This deuterated version is particularly useful as a tracer and internal standard in various analytical techniques.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.