Learn More
Bioworld Carnivorous Plant (Nepenthes) Basal Salt Mixture, 50 L
Catalog No. 50254743
Encompass_Preferred
Carnivorous Plant (Nepenthes) Basal Salt MixtureContains Micronutrients, Macronutrients, and Vitamins required for Nepenthes growthSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 0.65 g/LSoluble in waterCommonly supplemented with MS Vitamins and Sucrose
Catalog No. 50-254-743 Supplier Bioworld Supplier No. 306300893
Carnivorous Plant (Nepenthes) Basal Salt MixtureContains Micronutrients, Macronutrients, and Vitamins required for Nepenthes growthSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 0.65 g/LSoluble in waterCommonly supplemented with MS Vitamins and Sucrose