Learn More
Medchemexpress LLC HY-N0644 10mg Medchemexpress, Carnosic acid CAS:3650-09-7 Purity:>98%
SDP
Catalog No. 501933655
Encompass_Preferred
Medchemexpress, HY-N0644 10mg Carnosic acid CAS:3650-09-7 Carnosic acid has demonstrated inhibition of oxidative stress and inflammation, suppression of cell proliferation, and antibacterial activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3655 Supplier Medchemexpress LLC Supplier No. HYN064410MG
Medchemexpress, HY-N0644 10mg Carnosic acid CAS:3650-09-7 Carnosic acid has demonstrated inhibition of oxidative stress and inflammation, suppression of cell proliferation, and antibacterial activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.