Learn More
Enzo Life Sciences Carnosic acid, (10mg), CAS Number: 3650-09-7
SDP
Supplier:  Enzo Life Sciences ALX270264M010
Encompass_Preferred
PPARγ activator. Antioxidant. Naturally occurring phenolic compound with antioxidant properties. Inhibits lipid peroxidation induced by NADH or NADPH oxidation. Peroxisome proliferator-activated receptor γ (PPARγ) activator. Anti-inflammatory. Antimicrobial. Formula: C20H28O4. MW: 332.4. CAS Number: 3650-09-7. Purity: ≥95% (HPLC). Appearance: Yellow powder. Long Term Storage: -20°C
Catalog No. NC0143743