Learn More
Medchemexpress LLC Carnosic acid | 3650-09-7 | 332.43 | 5 MG
SDP
Catalog No. 5000389103
Encompass_Preferred
Carnosic acid is an orally active lipid absorption inhibitor, found abundantly in plants like *Rosmarinus officinalis L.* and *Salvia officinalis*. It inhibits oxidative stress and inflammation, suppresses cell proliferation, and exhibits antibacterial activity. It has therapeutic potential in various types of cancer and neuroprotective effects, mainly by activating the antioxidant NRF2/ARE pathway.
  • Inhibits lipid absorption
  • Reduces oxidative stress and inflammation
  • Suppresses cell proliferation
  • Exhibits antibacterial activity
  • Shows neuroprotective effects
Catalog No. 50-003-89103 Supplier Medchemexpress LLC Supplier No. HYN06445MG
Add to Cart
Add to Cart
Carnosic acid is an orally active lipid absorption inhibitor, found abundantly in plants like *Rosmarinus officinalis L.* and *Salvia officinalis*. It inhibits oxidative stress and inflammation, suppresses cell proliferation, and exhibits antibacterial activity. It has therapeutic potential in various types of cancer and neuroprotective effects, mainly by activating the antioxidant NRF2/ARE pathway.
  • Inhibits lipid absorption
  • Reduces oxidative stress and inflammation
  • Suppresses cell proliferation
  • Exhibits antibacterial activity
  • Shows neuroprotective effects