Learn More
Cayman Chemical Carnosic Acid, Purity: ≥95%, CAS: 3650-09-7, 50 mg
SDP
Catalog No. 502894015
Encompass_Preferred
An antioxidant derived from rosemary and common sage that inhibits lipid peroxidation and scavenges peroxyl radicals; cytotoxic towards various human cancer cell lines, reducing cell viability to 13-30% at 6.25 µg/ml
Catalog No. 50-289-4015 Supplier Cayman Chemical Supplier No. 8982050 MG
An antioxidant derived from rosemary and common sage that inhibits lipid peroxidation and scavenges peroxyl radicals; cytotoxic towards various human cancer cell lines, reducing cell viability to 13-30% at 6.25 µg/ml